Learn More
Thomas Scientific MyBlock Mini Digital Dry Bath, with cooling, 100-240V, US plug
SDP
Catalog No. 501458628
Encompass_Preferred
With its compact construction, broad temperature range, and multiple block options, the myBlock Mini is the perfect tool for laboratory incubations. The simple touch pad controls and a digital display provide for "set and walk away" temperature selection and unrivaled accuracy. While perfect for bench use, the optional 12V adapter expands the utility of the myBlock Mini to field use. At less than 4.3 inches wide, myBlock Mini is truly the first personal block incubator. Compact, fits on any crowded benchtop Available in heat only or heat & cool versions Exchangeable blocks for 0.2 to 50ml tubes Simple touchpad, digital control Link two consecutive programs (cooling model only) Custom blocks available upon request
Catalog No. 50-145-8628 Supplier Thomas Scientific Supplier No. 1190A99
With its compact construction, broad temperature range, and multiple block options, the myBlock Mini is the perfect tool for laboratory incubations. The simple touch pad controls and a digital display provide for "set and walk away" temperature selection and unrivaled accuracy. While perfect for bench use, the optional 12V adapter expands the utility of the myBlock Mini to field use. At less than 4.3 inches wide, myBlock Mini is truly the first personal block incubator. Compact, fits on any crowded benchtop Available in heat only or heat & cool versions Exchangeable blocks for 0.2 to 50ml tubes Simple touchpad, digital control Link two consecutive programs (cooling model only) Custom blocks available upon request