Learn More
Sigma Aldrich Fine Chemicals Biosciences MyBlockTM HL Mini dry bath1EA
Catalog No. 502905546
Encompass
The new MyBlock HL is the first and only mini heat block featuring a heated lid for protecting samples against unwanted condensation and evaporation.Both the block and lid temperature can be selected independently in increments of 0.1C making this MyBlock the ideal choice for the most temperature sensitive applications including immunoassays LAMP assays enzyme reactions other temperature hyper-sensitive procedures. Heated lid prevents evaporation condensationCompact fits in the palm of your handIndependant temperature control of lid blockExchangeable blocks for 0.2 to 0.5ml tubesRoom temperature to 37C in just 2 minutes
Catalog No. 50-290-5546 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7424941EA
The new MyBlock HL is the first and only mini heat block featuring a heated lid for protecting samples against unwanted condensation and evaporation.Both the block and lid temperature can be selected independently in increments of 0.1C making this MyBlock the ideal choice for the most temperature sensitive applications including immunoassays LAMP assays enzyme reactions other temperature hyper-sensitive procedures. Heated lid prevents evaporation condensationCompact fits in the palm of your handIndependant temperature control of lid blockExchangeable blocks for 0.2 to 0.5ml tubesRoom temperature to 37C in just 2 minutes