Learn More
Sigma Aldrich Fine Chemicals Biosciences MyBlockTM mini dry bath AC1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z6799841EA
Encompass
The MyBlock is truly the first personal dry bath block incubator both portable and affordableThe MyBlock is an impressive piece of equipment able to maintain temperatures from Room Temperature 5 to 100C and from between 0.1C increments to with in 0.2C uniformity throughout the samples. The controls are simple. You can just set it and walk away.Benchtop control of sample temperature has never been this easy or economical. The new MyBlock Mini fits almost anywhere and can even be used on-the-go in cars boats or wherever a 12 V power source is available. Measuring just 4.4 in. x 5.9 in. x 4.4 in. the MyBlock can fit in the palm of your hand.Supplied with clear cover and a block lifter the blocks themselves are sold separately to allow you the greatest versatility in temperature controlling your samples.Compact fits in the palm of your handExchangeable block for tubes 0.2 to 50 mL (blocks sold separately)Simple digital touch pad control designed for set and walk away t
Catalog No. 50-290-5672