Learn More
Aviva Systems Biology Corp MYC Antibody - N-terminal region (ARP30292_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30292_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MYC. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MYC. Alternate names: c-Myc, MRTL, bHLHe39ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-54147