Learn More
Aviva Systems Biology Corp MYC Antibody - C-terminal region (AVARP04001_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP04001_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MYC. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human MYC. Alternate names: MRTL, c-Myc, bHLHe39ApplicationWB. Species reactivity: Dog, Human, Mouse, Rat
Catalog No. 50-014-62531