Learn More
Description
Hybridization signals of digoxigenin-labeled polynucleotides appear as brown- to dark brown colored distinct dots (MYC gene region).
Normal situation: In interphases of normal cells or cells without an amplification involving the MYC gene locus, two distinct dot-shaped brown signals per nucleus appear.
Aberrant situation: In a cell with amplification of the MYC gene locus or polysomy of chromosome 8, multiple copies of the brown signal or brown signal clusters will be observed. Other signal distribution may be observed in some abnormal samples which might result in a different signal pattern than described above, indicating variant rearrangements.
Unexpected signal patterns should be further investigated.
Specifications
Specifications
| Content And Storage | The product is ready-to-use. No reconstitution, mixing, or dilution is required. Bring probe to room temperature (18 to 25°C) and mix briefly before use. |
| For Use With (Application) | CISH-P |
| Includes | Reagent Provided: 1. Digoxigenin-labeled polynucleotides targeting sequences mapping in 8q24.21 (chr8:128,596,776-128,887,929) harboring the MYC gene 2. Formamide based hybridization buffer |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.