Learn More
OriGene MYC/DDK Tagged Western Blot Molecular Weight Markers, 50 ug
SDP
Supplier:  OriGene MWM1001
Encompass_Preferred
Pre-mixed molecular weight markers with 6 over-expressed cell lysates with Myc/DDK tags covering 28 to 158 KDa. Each Vial contains 50ug total protein per tube; Sufficient for 20 loads.
Catalog No. NC0679914