Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Myc protein, human (C-terminal 6xHis) | >95.0% | 100UG
SDP

Catalog No. 5000174273
Encompass_Preferred

Recombinant full-length human MYC (c-Myc) protein expressed in Escherichia coli and supplied as a lyophilized powder with a C-terminal 6xHis tag. The product is formulated in PBS with trehalose for stability, tested for purity and endotoxin, and supplied with storage and reconstitution guidance for research use.

  • Species human.
  • Expression system Escherichia coli.
  • Tag C-terminal 6xHis.
  • Purity greater than 95% (SDS-PAGE).
  • Approximate molecular weight 58 kDa.
  • Lyophilized formulation from PBS, pH 7.4, with 8% trehalose.
  • Endotoxin level less than 1 EU/μg.
  • Store at -20°C; stable after reconstitution at 4°C for 1 week or -20°C for long term.

Catalog No. 50-001-74273 Supplier Medchemexpress LLC Supplier No. HYP704066100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant full-length human MYC (c-Myc) protein expressed in Escherichia coli and supplied as a lyophilized powder with a C-terminal 6xHis tag. The product is formulated in PBS with trehalose for stability, tested for purity and endotoxin, and supplied with storage and reconstitution guidance for research use.

  • Species human.
  • Expression system Escherichia coli.
  • Tag C-terminal 6xHis.
  • Purity greater than 95% (SDS-PAGE).
  • Approximate molecular weight 58 kDa.
  • Lyophilized formulation from PBS, pH 7.4, with 8% trehalose.
  • Endotoxin level less than 1 EU/μg.
  • Store at -20°C; stable after reconstitution at 4°C for 1 week or -20°C for long term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.