Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Myc protein, human (C-His) | 95.0% | 20UG
SDP

Catalog No. 5000174023
Encompass_Preferred

Recombinant human MYC protein with a C-terminal 6xHis tag, expressed in Escherichia coli and supplied lyophilized. Characterized by ≥95.0% purity and an approximate molecular weight of 58 kDa. Supplied lyophilized from PBS, pH 7.4 with 8% trehalose; intended for research use in biochemical and protein interaction studies.

  • Recombinant full-length human protein suitable for biochemical assays.
  • C-terminal 6xHis tag enables affinity purification and detection.
  • High purity (95.0%) confirmed by reducing SDS-PAGE.
  • Lyophilized formulation stabilized in PBS with 8% trehalose.
  • Recommended storage at -20°C; aliquot and freeze for long-term storage.
  • Do not reconstitute below 100 μg/mL in ddH2O.

Catalog No. 50-001-74023 Supplier Medchemexpress LLC Supplier No. HYP70406620UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human MYC protein with a C-terminal 6xHis tag, expressed in Escherichia coli and supplied lyophilized. Characterized by ≥95.0% purity and an approximate molecular weight of 58 kDa. Supplied lyophilized from PBS, pH 7.4 with 8% trehalose; intended for research use in biochemical and protein interaction studies.

  • Recombinant full-length human protein suitable for biochemical assays.
  • C-terminal 6xHis tag enables affinity purification and detection.
  • High purity (95.0%) confirmed by reducing SDS-PAGE.
  • Lyophilized formulation stabilized in PBS with 8% trehalose.
  • Recommended storage at -20°C; aliquot and freeze for long-term storage.
  • Do not reconstitute below 100 μg/mL in ddH2O.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.