Learn More
Proteintech Group Inc Myc-Trap Agarose, kit
SDP
Catalog No. 502258940 Shop All Proteintech Group Inc Products
Encompass
Myc-Trap Agarose is an affinity reagent for IP and purification of Myc-tagged proteins. It consists of a Myc Nanobody/ VHH coupled to agarose beads.
This item is not returnable. View return policy
Catalog No. 50-225-8940 Supplier Proteintech Group Inc Supplier No. YTAK20
Myc-Trap Agarose is an affinity reagent for IP and purification of Myc-tagged proteins. It consists of a Myc Nanobody/ VHH coupled to agarose beads.