Learn More
Proteintech Group Inc Myc-Trap Magnetic Agarose, kit
SDP
Supplier:  Proteintech Group Inc YTMAK20
Encompass
The ChromoTek Myc-Trap Magnetic Agarose is an affinity resin for IP of Myc-fusion proteins. It consists of a Myc Nanobody/VHH coupled to magnetic agarose beads.
Catalog No. 50-225-8947