Learn More
BIOHIPPO Myc-V5/GFP
SDP
Catalog No. 5000331883
Encompass
Myc-V5/GFP is a lentiviral vector expressing mouse myelocytomatosis oncogene Myc under the CMV promoter Each vial provides a nominal titer of 2 5x10E6 transducing units TU The construct contains a Myc ORF cDNA fused to a C terminal V5 tag together with a GFP reporter separated from the ORF by a self cleaving peptide to enable convenient monitoring of transduction and expression levels Lentiviral particles are supplied ready to use and are optimized for efficient gene delivery high gene expression and reliable genome integration in mammalian cells These particles allow easy transduction and stable overexpression of Myc and support establishment of long term reporter cell lines This product is intended for research use only and is not for diagnostic or therapeutic procedures
Catalog No. 50-003-31883 Supplier BIOHIPPO Supplier No. BHV194000971
Myc-V5/GFP is a lentiviral vector expressing mouse myelocytomatosis oncogene Myc under the CMV promoter Each vial provides a nominal titer of 2 5x10E6 transducing units TU The construct contains a Myc ORF cDNA fused to a C terminal V5 tag together with a GFP reporter separated from the ORF by a self cleaving peptide to enable convenient monitoring of transduction and expression levels Lentiviral particles are supplied ready to use and are optimized for efficient gene delivery high gene expression and reliable genome integration in mammalian cells These particles allow easy transduction and stable overexpression of Myc and support establishment of long term reporter cell lines This product is intended for research use only and is not for diagnostic or therapeutic procedures