Learn More
Cayman Chemical MycelIa sterIlIa MyrIocIn 10mg
SDP
Catalog No. 502892696
Encompass_Preferred
Myriocin is an amino fatty acid antibiotic derived from certain thermophylic fungi, in this case Mycelia sterilia. It is a potent immunosuppressant having 10- to 100-fold more activity than cyclosporin A.{8482} Myriocin is a potent inhibitor of serine palmitoyltransferase (Ki = 0.28 nM), the enzyme that catalyzes the first step of sphingolipid biosynthesis. {14764} It disrupts substratum adhesion of melanoma cells.{14763} It also suppresses cell proliferation in the murine cytotoxic T cell line CTLL-2 (IC50 = 15 nM) via apoptosis.{14764,2816} Myriocin suppresses replication of the hepatitis C virus (HCV) in a murine model.{14762}
Catalog No. 50-289-2696 Supplier Cayman Chemical Supplier No. 6315010 MG
Myriocin is an amino fatty acid antibiotic derived from certain thermophylic fungi, in this case Mycelia sterilia. It is a potent immunosuppressant having 10- to 100-fold more activity than cyclosporin A.{8482} Myriocin is a potent inhibitor of serine palmitoyltransferase (Ki = 0.28 nM), the enzyme that catalyzes the first step of sphingolipid biosynthesis. {14764} It disrupts substratum adhesion of melanoma cells.{14763} It also suppresses cell proliferation in the murine cytotoxic T cell line CTLL-2 (IC50 = 15 nM) via apoptosis.{14764,2816} Myriocin suppresses replication of the hepatitis C virus (HCV) in a murine model.{14762}