Learn More
Selleck Chemical LLC MYCi361 5mg
SDP
Supplier:  Selleck Chemical LLC S89055MG
Encompass_Preferred
MYCi361 is aMYC inhibitor that engages MYC inside cells, disrupts MYC/MAX dimers, and impairs MYC-driven gene expression. MYCi361 binds to MYC with Kd of 3.2 ?M. MYCi361 suppresses in vivo tumor growth, increases tumor immune cell infiltration, upregulates PD-L1 on tumors, and sensitizes tumors to anti-PD1 immunotherapy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3427