Learn More
Selleck Chemical LLC MYCi361 S8905-25MG
SDP
Catalog No. 5000024366
Encompass_Preferred
MYCi361 is a MYC inhibitor that engages MYC inside cells disrupts MYC/MAX dimers and impairs MYC-driven gene expression MYCi361 binds to MYC with Kd of 3 2 M MYCi361 suppresses in vivo tumor growth increases tumor immune cell infiltration upregulates PD-L1 on tumors and sensitizes tumors to anti-PD1 immunotherapy
Catalog No. 50-000-24366 Supplier Selleck Chemical LLC Supplier No. S890525MG
MYCi361 is a MYC inhibitor that engages MYC inside cells disrupts MYC/MAX dimers and impairs MYC-driven gene expression MYCi361 binds to MYC with Kd of 3 2 M MYCi361 suppresses in vivo tumor growth increases tumor immune cell infiltration upregulates PD-L1 on tumors and sensitizes tumors to anti-PD1 immunotherapy