Learn More
Selleck Chemical LLC MYCi975 S8906-10MM/1ML
SDP
Catalog No. 5000025244
Encompass_Preferred
MYCi975 (NUCC-0200975) is a potent selective and orally active inhibitor of MYC that disrupts MYC/MAX interaction promotes MYC T58 phosphorylation and MYC degradation and impairs MYC driven gene expression MYCi975 exhibits potent anti-tumor activities
Catalog No. 50-000-25244 Supplier Selleck Chemical LLC Supplier No. S890610MM/1ML
MYCi975 (NUCC-0200975) is a potent selective and orally active inhibitor of MYC that disrupts MYC/MAX interaction promotes MYC T58 phosphorylation and MYC degradation and impairs MYC driven gene expression MYCi975 exhibits potent anti-tumor activities