Learn More
Selleck Chemical LLC MYCi975-5mg
SDP
Supplier:  Selleck Chemical LLC S89065MG
Encompass_Preferred
MYCi975 (NUCC-0200975) is a potent, selective and orally active inhibitor of MYC that disrupts MYC/MAX interaction, promotes MYC T58 phosphorylation and MYC degradation, and impairs MYC driven gene expression. MYCi975 exhibits potent anti-tumor activities.
Catalog No. 50-238-9822