Learn More
Medchemexpress LLC HY-B2148 100mg Medchemexpress, Myclobutanil CAS:88671-89-0 Purity:>98%
SDP
Catalog No. 501933084
Encompass_Preferred
Medchemexpress, HY-B2148 100mg Myclobutanil CAS:88671-89-0 Myclobutanil is a conazole class fungicide widely used as an agrichemical. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3084 Supplier Medchemexpress LLC Supplier No. HYB2148100MG
Medchemexpress, HY-B2148 100mg Myclobutanil CAS:88671-89-0 Myclobutanil is a conazole class fungicide widely used as an agrichemical. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.