Learn More
Apexbio Technology LLC MYCLOBUTANIL-200MG
SDP
Catalog No. 5000568439
Encompass_Preferred
Myclobutanil (CAS No 88671-89-0) is a synthetic fungicide primarily studied in agricultural and environmental research It is an organo-sulfur compound originating from the class of triazole fungicides known for inhibiting the biosynthesis of ergosterol a critical component of fungal cell membranes Myclobutanil acts by targeting the cytochrome P450 enzyme pathway specifically the lanosterol 14 -demethylase disrupting cellular homeostasis in fungi Research frequently observes in vitro activity in the nanomolar to low micromolar range making it effective for antifungal applications Typically myclobutanil is employed in studies involving plant cell cultures and environmental impact assessments Additionally it serves as a model compound in drug screening assays Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-68439 Supplier Apexbio Technology LLC Supplier No. C6981200
Myclobutanil (CAS No 88671-89-0) is a synthetic fungicide primarily studied in agricultural and environmental research It is an organo-sulfur compound originating from the class of triazole fungicides known for inhibiting the biosynthesis of ergosterol a critical component of fungal cell membranes Myclobutanil acts by targeting the cytochrome P450 enzyme pathway specifically the lanosterol 14 -demethylase disrupting cellular homeostasis in fungi Research frequently observes in vitro activity in the nanomolar to low micromolar range making it effective for antifungal applications Typically myclobutanil is employed in studies involving plant cell cultures and environmental impact assessments Additionally it serves as a model compound in drug screening assays Experimental concentrations typically depend on the specific research design