Learn More
Cayman Chemical MyclobutnIl 50mg
SDP
Catalog No. 502933311
Encompass_Preferred
An analytical reference standard categorized as a triazole fungicide; found as a contaminant in concentrated Cannabis extracts from the California medical Cannabis market; intended for research and forensic applications
Catalog No. 50-293-3311 Supplier Cayman Chemical Supplier No. 2410050 MG
An analytical reference standard categorized as a triazole fungicide; found as a contaminant in concentrated Cannabis extracts from the California medical Cannabis market; intended for research and forensic applications