Learn More
Medchemexpress LLC HY-124675 5mg Medchemexpress, MYCMI-6 CAS:681282-09-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1246755MG
Encompass_Preferred
Medchemexpress, HY-124675 5mg MYCMI-6 CAS:681282-09-7 MYCMI-6 (NSC354961) is a potent and selective endogenous MYC:MAX protein interactions inhibitor. MYCMI-6 blocks MYC-driven transcription and binds selectively to the MYC bHLHZip domain with a Kd of 1.6 μM. MYCMI-6 inhibits tumor cell growth in a MYC-dependent manner (IC50<0.5 μM). MYCMI-6 is not cytotoxic to normal human cells. MYCMI-6 induces apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0000