Learn More
Abnova™ MYCN CISH Probe
Catalog No. 89335569 Shop All Abnova Corporation Products
Change view
Click to view available options
For Use With (Application):
CISH-P
Catalog No. For Use With (Application)
89-335-569 CISH-P
1 options
Catalog No. 89-335-569 Supplier Abnova™ Supplier No. CG0022
Add to Cart
Add to Cart
MYCN CISH Probe is designed for the qualitative detection human MYCN gene in formalin-fixed, paraffin-embedded tissue or cells by chromogenic in situ hybridization (CISH).
The probe is intended to be used in combination with the CISH Implementation Kit 2, which provides necessary reagents for specimen pretreatment and post-hybridization processing.
Hybridization signals of digoxigenin-labeled polynucleotides appear as brown- to dark brown colored distinct dots (MYCN gene).
Normal situation: In interphases of normal cells or cells without an amplification involving the MYCN gene locus, two distinct dot-shaped brown signals per nucleus appear.
Aberrant situation: In a cell with amplification of the MYCN gene locus or aneuploidy of chromosome 2, multiple copies of the brown signal or brown signal clusters will be observed.
Specifications
Content And Storage The product is ready-to-use. No reconstitution, mixing, or dilution is required. Bring probe to room temperature (18 to 25°C) and mix briefly before use.
For Use With (Application) CISH-P
Includes Reagent Provided:
Digoxigenin-labeled oligonucleotides targeting sequences mapping in 2p24.3* (chr2:15,846,046-16,213,717) harboring the MYCN gene.
2. Formamide based hybridization buffer.
*According to Human Genome Assembly GRCh37/hg19