Learn More
Aviva Systems Biology Corp MYCNOS Antibody - N-terminal region (ARP51638_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51638_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MYCNOS. It was validated on Western Blot. . Alternate names: NCYM, NYCM, MYCN-AS1ApplicationWB. Species reactivity: Human
Catalog No. 50-285-582