Learn More
IMMY Myco DDR™ Mycobacterial Digestion/ Decontamination Kits and Components
SDP
Catalog No. 22122328
Click to view available options
Type:
NaOH/NALC 2.5% Bulk
NaOH/NALC 3.0% Bulk
Neutralization Buffer B
Resuspension Buffer C
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
22-122-328 Neutralization Buffer B
22-122-326 NaOH/NALC 3.0% Bulk
22-122-325 NaOH/NALC 2.5% Bulk
22-122-327 Resuspension Buffer C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-122-328 Supplier IMMY Supplier No. TBPN6760
This item is not returnable. View return policy
Empowers labs to maximize the retention of viable Mycobacteria via rigid pH control throughout the digestion/decontamination process
  • Culture results obtained faster
  • Fewer specimens to reprocess
  • Reduced risk of cross-contamination
  • Easy to process specimens containing blood
  • Reduces sample splitting workload
  • Eliminates user variability, reducing the chances for technical error and allowing samples to be easily processed
  • Advanced buffer reduces the number of samples that need to be split
  • Cleaner smears
  • Visual result confirmation
Specifications
Type Neutralization Buffer B
For Use With (Application) Mycobacterial testing
Quantity 60 x 30 mL