Learn More
Myco Medical GLASSVAN™ Sterile Steel Blades
SDP
Catalog No. 22999111
Click to view available options
Size:
# 22
# 60
# 70
Blade Material:
Carbon Steel
Stainless Steel
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Blade Material
22-999-111 # 22 Stainless Steel
22-999-110 # 60 Stainless Steel
22-999-112 # 22 Carbon Steel
22-999-113 # 70 Carbon Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22-999-111 Supplier Myco Medical Supplier No. 3001T22
Computerized grinding and proprietary polishing/deburring processes ensure consistent, clean, sharp cutting edges
  • Available as carbon steel or stainless steel blades
  • VCI strips ensure product and packaging integrity
Specifications
Blade Material Stainless Steel
Product Type Steel blade
Size # 22
Sterility Sterile
Quantity Pk. of 10