Learn More
AbD Serotec Mycoplasma Removal Agent, 5 mL
Catalog No. NC9369822
Encompass
Mycoplasma Removal Agent; Developed for cell culture; 5mL; Non toxic; Prevent initial infection of cells in culture by mycoplasma; Cytotoxicity of MRA is low and cell toxicity is rare; Stable at RT
Catalog No. NC9369822 Supplier AbD Serotec Supplier No. BUF035
Mycoplasma Removal Agent; Developed for cell culture; 5mL; Non toxic; Prevent initial infection of cells in culture by mycoplasma; Cytotoxicity of MRA is low and cell toxicity is rare; Stable at RT