Learn More
Lonza Walkersville MycoAlert Mycoplasma Detection Kit (50 Tests)
Catalog No. NC9922140
Encompass
Lonza’s MycoAlert Mycoplasma Detection Assays are selective biochemical tests to detect Mycoplasma contamination in cell cultures. They exploit the activity of mycoplasmal enzymes which are found in the vast majority of about 200 mycoplasma species, but are not present in eukaryotic cells. Benefits:Convenient and fastSimply add two reagents to the culture supernatant Perform two luminescence readingsResults in less than 20 minutesDetects all common mollicute contaminations (Mycoplasma, Acholeplasma, Entomoplasma and Spiroplasma except Ureaplasma)Negligible interference with media componentsEasy interpretation of resultsContent:2 x 2.5 mL MycoAlert Reagent (lyophilized)2 x 2.5 mL MycoAlert Substrate (lyophilized)1 x 10 mL MycoAlert Assay Buffer
Catalog No. NC9922140 Supplier Lonza Walkersville Supplier No. LT07418
Lonza’s MycoAlert Mycoplasma Detection Assays are selective biochemical tests to detect Mycoplasma contamination in cell cultures. They exploit the activity of mycoplasmal enzymes which are found in the vast majority of about 200 mycoplasma species, but are not present in eukaryotic cells. Benefits:Convenient and fastSimply add two reagents to the culture supernatant Perform two luminescence readingsResults in less than 20 minutesDetects all common mollicute contaminations (Mycoplasma, Acholeplasma, Entomoplasma and Spiroplasma except Ureaplasma)Negligible interference with media componentsEasy interpretation of resultsContent:2 x 2.5 mL MycoAlert Reagent (lyophilized)2 x 2.5 mL MycoAlert Substrate (lyophilized)1 x 10 mL MycoAlert Assay Buffer