Learn More
Lonza Walkersville MycoAlert Mycoplasma Detection Kit (25 Tests)
Catalog No. NC9719283
Encompass
Lonza’s MycoAlert Mycoplasma Detection Assays are selective biochemical tests to detect Mycoplasma contamination in cell cultures. They exploit the activity of mycoplasmal enzymes which are found in the vast majority of about 200 mycoplasma species, but are not present in eukaryotic cells. Benefits:Convenient and fastSimply add two reagents to the culture supernatantPerform two luminescence readingsResults in less than 20 minEasy interpretation of results Detects all common mollicute contaminations (Mycoplasma, Acholeplasma, Entomoplasma and Spiroplasma except Ureaplasma)Negligible interference with media componentsContent:5 x 600 µL MycoAlert Reagent (lyophilized)5 x 600 µL MycoAlert Substrate (lyophilized)1 x 10 mL MycoAlert Assay Buffer
Catalog No. NC9719283 Supplier Lonza Walkersville Supplier No. LT07218
Lonza’s MycoAlert Mycoplasma Detection Assays are selective biochemical tests to detect Mycoplasma contamination in cell cultures. They exploit the activity of mycoplasmal enzymes which are found in the vast majority of about 200 mycoplasma species, but are not present in eukaryotic cells. Benefits:Convenient and fastSimply add two reagents to the culture supernatantPerform two luminescence readingsResults in less than 20 minEasy interpretation of results Detects all common mollicute contaminations (Mycoplasma, Acholeplasma, Entomoplasma and Spiroplasma except Ureaplasma)Negligible interference with media componentsContent:5 x 600 µL MycoAlert Reagent (lyophilized)5 x 600 µL MycoAlert Substrate (lyophilized)1 x 10 mL MycoAlert Assay Buffer