Learn More
ATCC Acetivibrio thermocellus; genomic DNA
Supplier:  ATCC 27405D5
Encompass
Genomic DNA isolated from Acetivibrio thermocellus strain VPI 7372 (ATCC 27405). This product can be used in PCR and other molecular biology applications.
Catalog No. 50-238-0570