Learn More
ATCC Mycobacterium avium
Catalog No. 502382257
Encompass
Standard strain for MAC susceptibility testing.
Catalog No. 50-238-2257 Supplier ATCC Supplier No. 700898
Add to Cart
Add to Cart
Standard strain for MAC susceptibility testing.