Learn More
ATCC Bacillus sp.
Supplier:  ATCC 700872
Encompass
This strain of Bacillus sp. was isolated from soil. It is cited to produce high levels of degradation enzymes but does not produce B. cereus-like endotoxin.
Catalog No. 50-237-9958