Learn More
ATCC Mycobacterium fortuitum; subsp. fortuitum
Supplier:  ATCC 6841
Encompass
Mycobacterium fortuitum subsp. fortuitum strain [TMC 1529] is a whole-genome sequenced bacterial type strain that is isolated from a cold abscess. This culture can be used in media testing and it produces restriction endonuclease MfoAI.
Catalog No. 50-238-4505