Learn More
ATCC Mycobacterium kansasii
Supplier:  ATCC 12478
Encompass
Mycobacterium kansasii strain G133 Bostrom [TMC 1204] is a whole-genome sequenced bacterial type strain. The culture was isolated from a fatal case and has applications in bioinformatics, media testing, and drug susceptibility testing.
Catalog No. 50-238-1787