Learn More
ATCC Herminiimonas species
Catalog No. 502380426
Encompass
Used in the assay of assimilable organic carbon.
Catalog No. 50-238-0426 Supplier ATCC Supplier No. 49643
Used in the assay of assimilable organic carbon.