Learn More
ATCC Mycobacterium peregrinum
Catalog No. 502384736
Encompass
Mycobacterium peregrinum strain MF1353 [CDC 97-1188] was isolated in Greenville, North Carolina, from the sputum of an immunocompromised patient. This whole-genome sequenced bacterial strain has applications in susceptibility disc testing.
Catalog No. 50-238-4736 Supplier ATCC Supplier No. 700686
Mycobacterium peregrinum strain MF1353 [CDC 97-1188] was isolated in Greenville, North Carolina, from the sputum of an immunocompromised patient. This whole-genome sequenced bacterial strain has applications in susceptibility disc testing.