Learn More
ATCC Mycobacterium smegmatis
Supplier:  ATCC 607
Encompass
Mycobacterium smegmatis was isolated from a human specimen. This bacterium is recommended as a test microorganism for USP <81> and can be used as a bacteriophage host.
Catalog No. 50-238-1758