Learn More
ATCC Mycobacterium smegmatis
Catalog No. 502381826
Encompass
Mycobacterium smegmatis strain mc(2)155 was derived from an existing strain (- mc(2)154) that was grown in the absence of kanamycin. This whole-genome sequenced bacterial strain is sensitive to kanamycin and can be used as a transformation host.
Catalog No. 50-238-1826 Supplier ATCC Supplier No. 700084
Add to Cart
Add to Cart
Mycobacterium smegmatis strain mc(2)155 was derived from an existing strain (- mc(2)154) that was grown in the absence of kanamycin. This whole-genome sequenced bacterial strain is sensitive to kanamycin and can be used as a transformation host.