Learn More
ATCC Streptococcus agalactiae; Strain: 2008232738
Catalog No. 502380173
Encompass
Sensitive to Ampicillin, Levofloxacin, Linezolid, Vancomycin, and Tigecycline. Resistant to Tetracycline.
Catalog No. 50-238-0173 Supplier ATCC Supplier No. BAA2675
Sensitive to Ampicillin, Levofloxacin, Linezolid, Vancomycin, and Tigecycline. Resistant to Tetracycline.