Learn More
ATCC Mycobacterium terrae; Strain W-45
Catalog No. 502384880
Encompass
Mycobacterium terrae strain W-45 is a whole-genome sequenced bacterial type strain. It has applications in efficacy testing, testing bactericides, and testing mycobactericides.
Catalog No. 50-238-4880 Supplier ATCC Supplier No. 15755
Mycobacterium terrae strain W-45 is a whole-genome sequenced bacterial type strain. It has applications in efficacy testing, testing bactericides, and testing mycobactericides.