Learn More
ATCC Mycobacterium tuberculosis; genomic DNA
Catalog No. 502382781
Encompass
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
Genomic DNA from Mycobacterium tuberculosis strain H37Ra (ATCC 25177). This whole-genome sequenced product can be used in PCR and other molecular biology applications, infectious disease research, and respiratory disease research.
Catalog No. 50-238-2781 Supplier ATCC Supplier No. 25177D5
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
Genomic DNA from Mycobacterium tuberculosis strain H37Ra (ATCC 25177). This whole-genome sequenced product can be used in PCR and other molecular biology applications, infectious disease research, and respiratory disease research.