Learn More
ATCC Mycobacterium tuberculosis; Quantitative DNA
Catalog No. 502381769
Encompass
Quantitative Genomic DNA from Mycobacterium tuberculosis strain H37Ra (ATCC 25177) can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load and for other molecular biology applications.
Catalog No. 50-238-1769 Supplier ATCC Supplier No. 25177DQ
Quantitative Genomic DNA from Mycobacterium tuberculosis strain H37Ra (ATCC 25177) can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load and for other molecular biology applications.