Learn More
ATCC Mycobacterium tuberculosis variant bovis BCG
Catalog No. 502382779
Encompass
Mycobacterium tuberculosis variant bovis BCG strain TMC 1010 [BCG Danish] is a whole-genome sequenced bacterium that was isolated from bovine milk. This strain exhibits low virulence in mice and guinea pigs.
Catalog No. 50-238-2779 Supplier ATCC Supplier No. 35733
Mycobacterium tuberculosis variant bovis BCG strain TMC 1010 [BCG Danish] is a whole-genome sequenced bacterium that was isolated from bovine milk. This strain exhibits low virulence in mice and guinea pigs.