Learn More
ATCC Mycobiome Whole; Cell Mix
Catalog No. 502383123
Encompass
NGS Standards are mock microbial communities that mimic mixed metagenomic samples. This product comprises an even mixture of 10 fully sequenced, characterized, and authenticated fungal strains selected on the basis of genus classification, genomic size, and other special features. Each order for this product is provided with access to One Codex, the leading bioinformatics platform for microbial genomics and metagenomics.
Catalog No. 50-238-3123 Supplier ATCC Supplier No. MSA2010
NGS Standards are mock microbial communities that mimic mixed metagenomic samples. This product comprises an even mixture of 10 fully sequenced, characterized, and authenticated fungal strains selected on the basis of genus classification, genomic size, and other special features. Each order for this product is provided with access to One Codex, the leading bioinformatics platform for microbial genomics and metagenomics.