Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MycoFluor™ Mycoplasma Detection Kit

Catalog No. m7006
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M7006 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M7006 Supplier Invitrogen™ Supplier No. M7006
Only null left
Add to Cart
Edge
Add to Cart

Identify mycoplasma infection in cell cultures visually

The MycoFluor™ Mycoplasma Detection Kit provides an ultrasensitive, rapid, and simple fluorescence microscopic assay for the visual identification of mycoplasma infection in laboratory cell cultures. In order to detect mycoplasma, the fluorescent MycoFluor reagent is added directly to the culture medium, with or without cells, and the stained sample is then examined under a fluorescence microscope.

Bioproduction, Cell Analysis, Cell Culture, Cell Tracing and Tracking, Clinical, Contaminant and Impurity QC Testing, Infectious Disease Diagnostics, Mammalian Cell Culture, Microbial Tracking, Molecular Diagnostics, Mycoplasma Detection

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2–8°C) and protect from light.
Detection Method Fluorescence
Form Liquid
Product Type Mycoplasma Detection
For Use With Fluorescence Microscope
Quantity 100 Tests
Shipping Condition Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.