Learn More
ATCC Escherichia coli; Strain: W3110
Supplier:  ATCC BAA3208
Encompass
Escherichia coli strain W3110 (ATCC BAA-3208) is an ATCC-manufactured and accessioned progeny of Escherichia coli (ATCC 39936) cited in US Patent Number 4,861,868. This strain can be used as a transformation host for the production of heterologous polypeptides that have N-terminal alanines.
Catalog No. 50-238-1595