Learn More
Aat Bioquest MycoLight Flow Cytometric Live Bacteria Assay Kit (100 Tests)
SDP
Catalog No. 502250070
Encompass_Preferred
The MycoLight Flow Cytometric Live Bacteria Assay Kit provides an easy and convenient methodfor evaluating bacterial vitality as a function of the intracellular esterase activity. This catalog is for 100 Tests.
This item is not returnable. View return policy
Catalog No. 50-225-0070 Supplier Aat Bioquest Supplier No. 22407
The MycoLight Flow Cytometric Live Bacteria Assay Kit provides an easy and convenient methodfor evaluating bacterial vitality as a function of the intracellular esterase activity. This catalog is for 100 Tests.