Learn More
Bioworld Mycological Agar, 500 g
Catalog No. 50254473
Encompass_Preferred
Mycological AgarUsed for the cultivation and maintenance of fungi, for the demonstration of chromogenesis, and for obtaining yeast and mold countspH: 7.0Dehydrated Appearance: light beige, homogeneous powderPrepared Appearance: light to medium amber agarSoluble in boiling waterComposition:Papaic Digest of Soybean Meal Dextrose Agar
Catalog No. 50-254-473 Supplier Bioworld Supplier No. 306260031
Mycological AgarUsed for the cultivation and maintenance of fungi, for the demonstration of chromogenesis, and for obtaining yeast and mold countspH: 7.0Dehydrated Appearance: light beige, homogeneous powderPrepared Appearance: light to medium amber agarSoluble in boiling waterComposition:Papaic Digest of Soybean Meal Dextrose Agar