Learn More
StatLab Mycoperm Blue Combination Stain and Coverslip for Mycology
SDP
Catalog No. 22130487
Change view
Click to view available options
Product Type:
Mycoperm Blue
Catalog No. Product Type
22-130-487 Mycoperm Blue
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-130-487 Supplier StatLab Supplier No. 753
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Keep accurate records by retaining mycology preparations for extended times with MycoPerm-BLUE, a combined stain/fixative
  • Liquid solution is packaged in a dropper bottle for easy application
  • Works as both a stain and fixative and makes messy nail polish obsolete
  • Tease mount preparations can be stored for long periods of time
  • Contains plasticizers which help make a durable, long-life preparation
  • 12 mL bottle
Specifications
Volume (Metric) 12 mL
Product Type Mycoperm Blue