Learn More
Cayman Chemical Mycophenolate Mofetil, Purity: ≥95%, CAS: 128794-94-5, 100 mg
SDP
Catalog No. 502955255
Encompass_Preferred
A prodrug form of mycophenolic acid; inhibits thymidine incorporation in primary rat aortic smooth muscle cells at 1 and 10 µg/ml; increases the duration of transplant motor activity in a rabbit model of retroperitoneal heterotopic heart transplantation at 12.5 mg/kg; eliminates formation of microbleeds and hemorrhages in the cerebrum of stroke-prone spontaneously hypertensive rats at 25 mg/kg; lowers MAP and reduces urinary albumin excretion and glomerulosclerosis in a mouse model of SLE at 60 mg/kg
Catalog No. 50-295-5255 Supplier Cayman Chemical Supplier No. 13988100 MG
A prodrug form of mycophenolic acid; inhibits thymidine incorporation in primary rat aortic smooth muscle cells at 1 and 10 µg/ml; increases the duration of transplant motor activity in a rabbit model of retroperitoneal heterotopic heart transplantation at 12.5 mg/kg; eliminates formation of microbleeds and hemorrhages in the cerebrum of stroke-prone spontaneously hypertensive rats at 25 mg/kg; lowers MAP and reduces urinary albumin excretion and glomerulosclerosis in a mouse model of SLE at 60 mg/kg