Learn More
Apexbio Technology LLC Mycophenolate Mofetil 128794-94-5 200mg
SDP
Catalog No. 5000095379
Encompass_Preferred
Mycophenolate Mofetil (128794-94-5) is a small-molecule inhibitor targeting inosine monophosphate dehydrogenase (IMPDH) isoforms I and II It is designed to selectively and reversibly inhibit IMPDH thereby disrupting de novo guanosine nucleotide biosynthesis Mycophenolate Mofetil exerts its biological activity primarily through non-competitive inhibition of IMPDH blocking the conversion of inosine monophosphate (IMP) to xanthosine monophosphate (XMP) In in vitro studies Mycophenolate Mofetil demonstrates inhibitory activity with IC50 values of 39 nM and 27 nM for IMPDH I and II respectively Based on these pharmacological properties Mycophenolate Mofetil holds research potential in studies on immunology purine metabolism regulation cell proliferation transplant rejection and autoimmune disease models
Catalog No. 50-000-95379 Supplier Apexbio Technology LLC Supplier No. A4336200
Mycophenolate Mofetil (128794-94-5) is a small-molecule inhibitor targeting inosine monophosphate dehydrogenase (IMPDH) isoforms I and II It is designed to selectively and reversibly inhibit IMPDH thereby disrupting de novo guanosine nucleotide biosynthesis Mycophenolate Mofetil exerts its biological activity primarily through non-competitive inhibition of IMPDH blocking the conversion of inosine monophosphate (IMP) to xanthosine monophosphate (XMP) In in vitro studies Mycophenolate Mofetil demonstrates inhibitory activity with IC50 values of 39 nM and 27 nM for IMPDH I and II respectively Based on these pharmacological properties Mycophenolate Mofetil holds research potential in studies on immunology purine metabolism regulation cell proliferation transplant rejection and autoimmune disease models